Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATCGGGCCCAGCTCCTGGAACC[A/G]TCCAAATCGGCCTCGTCCAAAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602986 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRG2 - developmentally regulated GTP binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001388.4 | 271 | Silent Mutation | CCA,CCG | P,P 44 | NP_001379.1 | |
XM_005256499.2 | 271 | Silent Mutation | CCA,CCG | P,P 44 | XP_005256556.1 | |
XM_005256500.2 | 271 | UTR 5 | XP_005256557.1 | |||
XM_011523704.1 | 271 | Silent Mutation | CCA,CCG | P,P 44 | XP_011522006.1 | |
XM_011523705.1 | 271 | Silent Mutation | CCA,CCG | P,P 44 | XP_011522007.1 | |
XM_011523706.1 | 271 | UTR 5 | XP_011522008.1 | |||
XM_011523707.1 | 271 | UTR 5 | XP_011522009.1 | |||
XM_017024296.1 | 271 | UTR 5 | XP_016879785.1 | |||
XM_017024297.1 | 271 | UTR 5 | XP_016879786.1 |