Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGCGGGCATCGTGATAGCCGAG[A/G]CGCTACAGAACCAGCTAGCCTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611045 MIM: 605315 MIM: 611793 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
G6PC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
G6PC3 - glucose-6-phosphatase catalytic subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319945.1 | 256 | Missense Mutation | ACG,GCG | T,A 14 | NP_001306874.1 | |
NM_138387.3 | 256 | Missense Mutation | ACG,GCG | T,A 14 | NP_612396.1 | |
XM_011525473.2 | 256 | Intron | XP_011523775.1 | |||
XM_011525474.2 | 256 | UTR 5 | XP_011523776.1 | |||
XM_017025335.1 | 256 | Intron | XP_016880824.1 |
HDAC5 - histone deacetylase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LSM12 - LSM12 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |