Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTTCTCCGGGTCGTACAACCGC[G/T]TGCAGCTTTGGCTGGTCGGGACGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 152391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALOX12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALOX12 - arachidonate 12-lipoxygenase, 12S type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000697.2 | 117 | Missense Mutation | GTG,TTG | V,L 22 | NP_000688.2 | |
XM_011523780.2 | 117 | Missense Mutation | GTG,TTG | V,L 141 | XP_011522082.1 |
ALOX12-AS1 - ALOX12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |