Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAAGCTTACCAGCTGCTGCTGAC[C/T]GACCACACTGTGACCTTCCTGCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612185 MIM: 608755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASKIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASKIN2 - CASK interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LLGL2 - LLGL2, scribble cell polarity complex component | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSEN54 - tRNA splicing endonuclease subunit 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207346.2 | 470 | Silent Mutation | ACC,ACT | T,T 146 | NP_997229.2 | |
XM_005257229.3 | 470 | Silent Mutation | ACC,ACT | T,T 146 | XP_005257286.1 |