Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATATAGTTAAAGATACCTGAACCA[C/T]AGAATATGGCAACCACAGTGAGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR3A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR3A1 - olfactory receptor family 3 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002550.2 | 764 | Missense Mutation | TAT,TGT | Y,C 255 | NP_002541.2 | |
XM_017024695.1 | 764 | Intron | XP_016880184.1 | |||
XM_017024696.1 | 764 | Intron | XP_016880185.1 |
OR3A2 - olfactory receptor family 3 subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002551.3 | 764 | Intron | NP_002542.3 | |||
XM_017024697.1 | 764 | Intron | XP_016880186.1 | |||
XM_017024698.1 | 764 | Intron | XP_016880187.1 |