Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGTTTGTGAAAGATGTTGCCACC[A/G]CGAGACATCAAGTCCAGCACAGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 146630 | ||||||||||||||||||||
Literature Links: |
ICAM2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ICAM2 - intercellular adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000873.3 | 2446 | Silent Mutation | CGC,CGT | R,R 199 | NP_000864.2 | |
NM_001099786.1 | 2446 | Silent Mutation | CGC,CGT | R,R 199 | NP_001093256.1 | |
NM_001099787.1 | 2446 | Silent Mutation | CGC,CGT | R,R 199 | NP_001093257.1 | |
NM_001099788.1 | 2446 | Silent Mutation | CGC,CGT | R,R 199 | NP_001093258.1 | |
NM_001099789.1 | 2446 | Silent Mutation | CGC,CGT | R,R 199 | NP_001093259.1 |
PRR29 - proline rich 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164257.1 | 2446 | UTR 3 | NP_001157729.1 | |||
NM_001191029.1 | 2446 | UTR 3 | NP_001177958.1 | |||
NM_001191030.1 | 2446 | UTR 3 | NP_001177959.1 | |||
NM_001191031.1 | 2446 | UTR 3 | NP_001177960.1 | |||
XM_005257805.4 | 2446 | Intron | XP_005257862.1 | |||
XM_011525468.2 | 2446 | Intron | XP_011523770.1 | |||
XM_011525469.1 | 2446 | UTR 3 | XP_011523771.1 | |||
XM_017025315.1 | 2446 | UTR 3 | XP_016880804.1 |
PRR29-AS1 - PRR29 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |