Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGTGGACAAAAAACTGGGACGCT[G/T]CTTCTTCTCTGGCTACGATAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114760 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CA4 - carbonic anhydrase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005257639.2 | 303 | Missense Mutation | TGC,TTC | C,F 70 | XP_005257696.1 | |
XM_011525183.2 | 303 | UTR 5 | XP_011523485.1 | |||
XM_017025012.1 | 303 | UTR 5 | XP_016880501.1 |