Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCAGCTGCTGAGCCAGTATGACA[A/G]CGAGAAGAAGTGGGAGCTCATCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604656 | ||||||||||||||||||||
Literature Links: |
FMNL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FMNL1 - formin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005892.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | NP_005883.2 | |
XM_006722062.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722125.1 | |
XM_006722063.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722126.1 | |
XM_006722064.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722127.1 | |
XM_006722065.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722128.1 | |
XM_006722066.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722129.1 | |
XM_006722069.2 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722132.1 | |
XM_006722070.3 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_006722133.1 | |
XM_011525179.2 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_011523481.1 | |
XM_011525180.2 | 575 | Missense Mutation | AAC,AGC | N,S 30 | XP_011523482.1 | |
XM_011525182.2 | 575 | Missense Mutation | AAC,AGC | N,S 61 | XP_011523484.1 |