Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTCGTAAAAATCTCCAGTTCAC[A/G]TAGAGCCTGTAAACATTCGGCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605532 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMURF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMURF2 - SMAD specific E3 ubiquitin protein ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022739.3 | 1743 | Silent Mutation | TAC,TAT | Y,Y 585 | NP_073576.1 | |
XM_005257585.3 | 1743 | Silent Mutation | TAC,TAT | Y,Y 572 | XP_005257642.1 | |
XM_011525128.1 | 1743 | Silent Mutation | TAC,TAT | Y,Y 587 | XP_011523430.1 |