Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGTGCACAGGCAGTCTCTCCCCTA[C/T]ATCCTTTCTGCGTCCTGCAGCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNS4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNS4 - tensin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032865.5 | 2385 | Missense Mutation | ATA,ATG | I,M 715 | NP_116254.4 | |
XM_005257744.1 | 2385 | Missense Mutation | ATA,ATG | I,M 714 | XP_005257801.1 | |
XM_017025236.1 | 2385 | Missense Mutation | ATA,ATG | I,M 689 | XP_016880725.1 | |
XM_017025237.1 | 2385 | Intron | XP_016880726.1 |