Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATTACTCTTCACTTATGACTT[C/T]TGTGTGAACCTCCCGACTCTTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 160742 | ||||||||||||||||||||
Literature Links: |
MYH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYH4 - myosin heavy chain 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017533.2 | 6055 | Missense Mutation | AAA,GAA | K,E 1934 | NP_060003.2 | |
XM_017024676.1 | 6055 | Missense Mutation | AAA,GAA | K,E 1934 | XP_016880165.1 |
MYHAS - myosin heavy chain gene cluster antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |