Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTGCCTGGTCTGTCGCTGCTTAA[A/G]TTTAGGGGGCCCTTTCCTGGGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180073 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6G PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE6G - phosphodiesterase 6G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002602.3 | 232 | Missense Mutation | CTT,TTT | L,F 30 | NP_002593.1 | |
XM_006721942.3 | 232 | Missense Mutation | CTT,TTT | L,F 80 | XP_006722005.1 | |
XM_017024734.1 | 232 | Missense Mutation | CTT,TTT | L,F 100 | XP_016880223.1 | |
XM_017024735.1 | 232 | Missense Mutation | CTT,TTT | L,F 30 | XP_016880224.1 | |
XM_017024736.1 | 232 | Missense Mutation | CTT,TTT | L,F 30 | XP_016880225.1 |
TSPAN10 - tetraspanin 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |