Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAACAACAACGAATGGACTCACC[A/G]ATGAACCCCATACTCCTTCCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108728 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACLY PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACLY - ATP citrate lyase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001096.2 | 3380 | Silent Mutation | ATC,ATT | I,I 1070 | NP_001087.2 | |
NM_001303274.1 | 3380 | Silent Mutation | ATC,ATT | I,I 1124 | NP_001290203.1 | |
NM_001303275.1 | 3380 | Silent Mutation | ATC,ATT | I,I 1114 | NP_001290204.1 | |
NM_198830.1 | 3380 | Silent Mutation | ATC,ATT | I,I 1060 | NP_942127.1 | |
XM_005257395.1 | 3380 | Silent Mutation | ATC,ATT | I,I 1070 | XP_005257452.1 | |
XM_017024688.1 | 3380 | Silent Mutation | ATC,ATT | I,I 1060 | XP_016880177.1 |
KLHL11 - kelch like family member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |