Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCCCCACCGGCCCAGCCGCCTGC[A/C]CCCCCGGCCAGCCAGCTCTCGGGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603460 MIM: 606539 MIM: 604449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK5R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK5R1 - cyclin dependent kinase 5 regulatory subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003885.2 | 532 | Silent Mutation | GCA,GCC | A,A 108 | NP_003876.1 | |
XM_011525438.2 | 532 | Silent Mutation | GCA,GCC | A,A 108 | XP_011523740.1 | |
XM_017025281.1 | 532 | Silent Mutation | GCA,GCC | A,A 108 | XP_016880770.1 |
MYO1D - myosin ID | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD11 - proteasome 26S subunit, non-ATPase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |