Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGGGAGCTGCCCGCCAACTGT[A/G]CCTGCAGGGTGGAAGAGAAATGAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611301 MIM: 606590 | ||||||||||||||||||||
Literature Links: |
FAAP100 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAAP100 - Fanconi anemia core complex associated protein 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPLOC4 - NPL4 homolog, ubiquitin recognition factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017921.3 | 1349 | Silent Mutation | AGC,AGT | S,S 557 | NP_060391.2 | |
XM_011524979.1 | 1349 | Intron | XP_011523281.1 | |||
XM_011524980.1 | 1349 | Missense Mutation | GCA,GTA | A,V 582 | XP_011523282.1 | |
XM_011524981.1 | 1349 | Silent Mutation | AGC,AGT | S,S 562 | XP_011523283.1 | |
XM_011524982.2 | 1349 | Intron | XP_011523284.1 | |||
XM_017024830.1 | 1349 | Missense Mutation | GCA,GTA | A,V 356 | XP_016880319.1 |