Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGCCGACAGGAAGACCACTGC[C/T]GGAGCTGCTCCTGGCTGAGGCAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600242 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCR7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCR7 - C-C motif chemokine receptor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301714.1 | 1139 | Missense Mutation | CAG,CGG | Q,R 290 | NP_001288643.1 | |
NM_001301716.1 | 1139 | Missense Mutation | CAG,CGG | Q,R 347 | NP_001288645.1 | |
NM_001301717.1 | 1139 | Missense Mutation | CAG,CGG | Q,R 347 | NP_001288646.1 | |
NM_001301718.1 | 1139 | Missense Mutation | CAG,CGG | Q,R 347 | NP_001288647.1 | |
NM_001838.3 | 1139 | Missense Mutation | CAG,CGG | Q,R 353 | NP_001829.1 |