Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTGACAACGCCCGGCCCCGCAC[C/T]GCCAGGGGAATGACTTTGCATACTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D28 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBC1D28 - TBC1 domain family member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039397.2 | Intron | NP_001034486.2 | ||||
XM_011523781.2 | Intron | XP_011522083.1 | ||||
XM_011523782.2 | Intron | XP_011522084.1 | ||||
XM_011523783.2 | Intron | XP_011522085.1 | ||||
XM_017024416.1 | Intron | XP_016879905.1 | ||||
XM_017024417.1 | Intron | XP_016879906.1 | ||||
XM_017024418.1 | Intron | XP_016879907.1 | ||||
XM_017024419.1 | Intron | XP_016879908.1 | ||||
XM_017024420.1 | Intron | XP_016879909.1 | ||||
XM_017024421.1 | Intron | XP_016879910.1 | ||||
XM_017024422.1 | Intron | XP_016879911.1 | ||||
XM_017024423.1 | Intron | XP_016879912.1 |