Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGGCTAGGAGAGACTGAAGTTCA[A/G]TTTCCAGGGTTTGAAGAGTGCGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616646 | ||||||||||||||||||||
Literature Links: |
KRT25 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRT25 - keratin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181534.3 | 981 | Missense Mutation | ACT,ATT | T,I 311 | NP_853512.1 | |
XM_011524414.1 | 981 | Missense Mutation | ACT,ATT | T,I 309 | XP_011522716.1 |