Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGTTCCAGAGAGCCCAGTCTCT[A/T]CTCAGCACCGACCGGGAGGCCTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604449 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMD11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMD11 - proteasome 26S subunit, non-ATPase 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270482.1 | 106 | Silent Mutation | CTA,CTT | L,L 15 | NP_001257411.1 | |
NM_002815.3 | 106 | Silent Mutation | CTA,CTT | L,L 15 | NP_002806.2 |