Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCTTGATCCCGCTGCAGAGAGT[C/T]ATATCCATCCTGTTTTTCTGCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRLF3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRLF3 - cytokine receptor like factor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015986.3 | 1313 | Missense Mutation | AAC,GAC | N,D 344 | NP_057070.3 | |
XM_011524879.1 | 1313 | Missense Mutation | AAC,GAC | N,D 228 | XP_011523181.1 |