Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGAGGTTTTCCAGATCAGTGACA[A/G]CCATGCTGGTTTGCTTCAGACTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611701 | ||||||||||||||||||||
Literature Links: |
SPNS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPNS3 - sphingolipid transporter 3 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320449.1 | 270 | UTR 5 | NP_001307378.1 | |||
NM_182538.4 | 270 | Missense Mutation | AAC,AGC | N,S 81 | NP_872344.3 | |
XM_011523723.1 | 270 | Missense Mutation | AAC,AGC | N,S 81 | XP_011522025.1 | |
XM_011523724.1 | 270 | Missense Mutation | AAC,AGC | N,S 81 | XP_011522026.1 | |
XM_011523725.1 | 270 | Missense Mutation | AAC,AGC | N,S 81 | XP_011522027.1 | |
XM_011523726.2 | 270 | Intron | XP_011522028.1 | |||
XM_011523728.2 | 270 | Intron | XP_011522030.1 | |||
XM_017024345.1 | 270 | Intron | XP_016879834.1 |