Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCAGATTTTTCCTCTGTTCCGTG[C/T]GCTAGGTGCAAACGGCCTCCTCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616864 | ||||||||||||||||||||
Literature Links: |
HEXDC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HEXDC - hexosaminidase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173620.2 | 244 | Missense Mutation | GCG,GTG | A,V 35 | NP_775891.2 | |
XM_005256346.3 | 244 | Intron | XP_005256403.1 | |||
XM_005256347.4 | 244 | Missense Mutation | GCG,GTG | A,V 35 | XP_005256404.1 | |
XM_006722280.3 | 244 | Intron | XP_006722343.1 | |||
XM_011523560.2 | 244 | Intron | XP_011521862.1 | |||
XM_011523561.2 | 244 | Intron | XP_011521863.1 | |||
XM_017024484.1 | 244 | Missense Mutation | GCG,GTG | A,V 35 | XP_016879973.1 | |
XM_017024485.1 | 244 | Intron | XP_016879974.1 |
OGFOD3 - 2-oxoglutarate and iron dependent oxygenase domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |