Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGAGGCTGTGCCCCTGGGCCTG[A/G]GCCTTGAGGATGCTGACTGGACCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615647 | ||||||||||||||||||||
Literature Links: |
TEX19 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX19 - testis expressed 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207459.3 | 800 | Missense Mutation | AGC,GGC | S,G 135 | NP_997342.1 | |
XM_017024638.1 | 800 | Missense Mutation | AGC,GGC | S,G 135 | XP_016880127.1 | |
XM_017024639.1 | 800 | Missense Mutation | AGC,GGC | S,G 135 | XP_016880128.1 |