Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTGTTTTCCTCCATGACTTTG[C/T]GCCTGTCTCGCTGCAGCTGCTCGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606735 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OSBPL7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OSBPL7 - oxysterol binding protein like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145798.2 | 2715 | Missense Mutation | CAC,CGC | H,R 791 | NP_665741.1 | |
XM_017024122.1 | 2715 | Missense Mutation | CAC,CGC | H,R 791 | XP_016879611.1 | |
XM_017024123.1 | 2715 | Missense Mutation | CAC,CGC | H,R 791 | XP_016879612.1 | |
XM_017024124.1 | 2715 | Missense Mutation | CAC,CGC | H,R 773 | XP_016879613.1 | |
XM_017024125.1 | 2715 | Missense Mutation | CAC,CGC | H,R 353 | XP_016879614.1 |