Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACATATTGAACAGAAACTCCTTC[C/T]CTAGCTCCAGCGTCTGGTATTTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 142963 MIM: 142964 MIM: 608632 | ||||||||||||||||||||
Literature Links: |
HOXB8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOXB8 - homeobox B8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024016.3 | 806 | Intron | NP_076921.1 | |||
XM_005257286.2 | 806 | Intron | XP_005257343.1 | |||
XM_017024564.1 | 806 | Intron | XP_016880053.1 |
HOXB9 - homeobox B9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024017.4 | 806 | Missense Mutation | GAG,GGG | E,G 201 | NP_076922.1 |
MIR196A1 - microRNA 196a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |