Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGCGCTCGCTAGAAAGCTGCTC[A/G]TTCACCTGGGTGGGCACAGAAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AATK-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AATK-AS1 - AATK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CEP131 - centrosomal protein 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009811.3 | 3202 | Silent Mutation | AAC,AAT | N,N 947 | NP_001009811.2 | |
NM_001319228.1 | 3202 | Silent Mutation | AAC,AAT | N,N 986 | NP_001306157.1 | |
NM_001319229.1 | 3202 | Silent Mutation | AAC,AAT | N,N 950 | NP_001306158.1 | |
NM_014984.3 | 3202 | Silent Mutation | AAC,AAT | N,N 983 | NP_055799.2 | |
XM_011524547.2 | 3202 | Silent Mutation | AAC,AAT | N,N 991 | XP_011522849.1 | |
XM_011524548.2 | 3202 | Silent Mutation | AAC,AAT | N,N 988 | XP_011522850.1 | |
XM_011524550.2 | 3202 | Silent Mutation | AAC,AAT | N,N 985 | XP_011522852.1 | |
XM_011524551.2 | 3202 | Silent Mutation | AAC,AAT | N,N 991 | XP_011522853.1 | |
XM_011524552.2 | 3202 | Silent Mutation | AAC,AAT | N,N 955 | XP_011522854.1 | |
XM_011524553.2 | 3202 | Silent Mutation | AAC,AAT | N,N 950 | XP_011522855.1 | |
XM_011524555.2 | 3202 | Silent Mutation | AAC,AAT | N,N 991 | XP_011522857.1 | |
XM_017024369.1 | 3202 | Silent Mutation | AAC,AAT | N,N 986 | XP_016879858.1 |