Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCGGTCATCTCTAGAAACTGCG[C/T]GTAGACATCTCGGATGCACTCCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609226 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
WDR45B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
WDR45B - WD repeat domain 45B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019613.3 | 1195 | Missense Mutation | ACG,GCG | T,A 334 | NP_062559.2 | |
XM_005256377.4 | 1195 | Intron | XP_005256434.1 | |||
XM_017024862.1 | 1195 | Intron | XP_016880351.1 | |||
XM_017024863.1 | 1195 | Intron | XP_016880352.1 |