Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGGTGAGCTTGGCACCTACATGA[C/G]ACCCGGCTTGGAGCACTCGCTGTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602494 | ||||||||||||||||||||
Literature Links: |
CCL23 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL23 - C-C motif chemokine ligand 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005064.5 | 369 | Missense Mutation | CTC,GTC | L,V 100 | NP_005055.3 | |
NM_145898.3 | 369 | Missense Mutation | CTC,GTC | L,V 83 | NP_665905.2 |