Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCATGAAACCAGACGGAGAAGCTG[C/T]GTCCATTCCACTGCAGGCAGCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611041 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM47 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM47 - tripartite motif containing 47 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033452.2 | 1607 | Missense Mutation | CAC,CGC | H,R 527 | NP_258411.2 | |
XM_005257787.4 | 1607 | Missense Mutation | CAC,CGC | H,R 289 | XP_005257844.1 | |
XM_005257788.4 | 1607 | Missense Mutation | CAC,CGC | H,R 289 | XP_005257845.1 |
TRIM65 - tripartite motif containing 65 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |