Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACAACACATGCAGCAATAATGG[A/G]CTCTATTGGTACACCCACAGTTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMIGD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMIGD1 - transmembrane and immunoglobulin domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319942.1 | 766 | Intron | NP_001306871.1 | |||
NM_206832.2 | 766 | Missense Mutation | CCC,TCC | P,S 223 | NP_996663.1 | |
XM_011524787.1 | 766 | Missense Mutation | CCC,TCC | P,S 223 | XP_011523089.1 |