Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCTACTTCCTGTACCAACTGCTG[C/T]GGGGCCTGAAGTACATGCACTCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614144 MIM: 602521 MIM: 600596 | ||||||||||||||||||||
Literature Links: |
B9D1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
B9D1 - B9 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MAPK7 - mitogen-activated protein kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002749.3 | 906 | Missense Mutation | CGG,TGG | R,W 168 | NP_002740.2 | |
NM_139032.2 | 906 | Missense Mutation | CGG,TGG | R,W 29 | NP_620601.1 | |
NM_139033.2 | 906 | Missense Mutation | CGG,TGG | R,W 168 | NP_620602.2 | |
NM_139034.2 | 906 | Missense Mutation | CGG,TGG | R,W 168 | NP_620603.2 | |
XM_006721557.3 | 906 | Missense Mutation | CGG,TGG | R,W 174 | XP_006721620.1 | |
XM_006721558.3 | 906 | Missense Mutation | CGG,TGG | R,W 174 | XP_006721621.1 | |
XM_006721559.3 | 906 | Missense Mutation | CGG,TGG | R,W 174 | XP_006721622.1 | |
XM_011523957.2 | 906 | Missense Mutation | CGG,TGG | R,W 29 | XP_011522259.1 |
MFAP4 - microfibrillar associated protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |