Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACAGAGCCACAGCAGGAGCTGA[A/C]CATGGCGTCAGAGGGTGGAGGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KRTAP4-2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KRTAP4-2 - keratin associated protein 4-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTAP4-3 - keratin associated protein 4-3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033187.1 | 4 | Missense Mutation | GTC,TTC | V,F 2 | NP_149443.1 |
KRTAP4-4 - keratin associated protein 4-4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |