Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGCAAGGGGGCTGCTCCCCCGCG[A/G]TGCCAGCCGCCCCCATGTGAGTTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601522 | ||||||||||||||||||||
Literature Links: |
GRB7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GRB7 - growth factor receptor bound protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030002.2 | 555 | Missense Mutation | GAT,GGT | D,G 97 | NP_001025173.1 | |
NM_001242442.1 | 555 | Missense Mutation | GAT,GGT | D,G 120 | NP_001229371.1 | |
NM_001242443.1 | 555 | Missense Mutation | GAT,GGT | D,G 97 | NP_001229372.1 | |
NM_005310.3 | 555 | Missense Mutation | GAT,GGT | D,G 97 | NP_005301.2 | |
XM_017024536.1 | 555 | UTR 5 | XP_016880025.1 | |||
XM_017024537.1 | 555 | Missense Mutation | GAT,GGT | D,G 97 | XP_016880026.1 | |
XM_017024538.1 | 555 | UTR 5 | XP_016880027.1 |