Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGGTCAAGAACTTCCGAGAGGCC[A/G]TGAACTGGACCCCATAATCCATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611221 MIM: 610075 | ||||||||||||||||||||
Literature Links: |
GSDMB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSDMB - gasdermin B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC3C - leucine rich repeat containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ORMDL3 - ORMDL sphingolipid biosynthesis regulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320801.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | NP_001307730.1 | |
NM_001320802.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | NP_001307731.1 | |
NM_001320803.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | NP_001307732.1 | |
NM_139280.3 | 322 | Missense Mutation | ACG,ATG | T,M 96 | NP_644809.1 | |
XM_017025345.1 | 322 | Missense Mutation | ACG,ATG | T,M 114 | XP_016880834.1 | |
XM_017025346.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | XP_016880835.1 | |
XM_017025347.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | XP_016880836.1 | |
XM_017025348.1 | 322 | Missense Mutation | ACG,ATG | T,M 96 | XP_016880837.1 |