Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACCTTGCTGTTGGTCCACGGGC[A/G]GGGCTTCCAAGGGACAGCAGCCAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608163 MIM: 603691 MIM: 604858 MIM: 610935 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC7 - exocyst complex component 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GALR2 - galanin receptor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRP68 - signal recognition particle 68 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZACN - zinc activated ion channel | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_180990.3 | 157 | Missense Mutation | CAG,CGG | Q,R 25 | NP_851321.2 |