Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTAAACCAATTGTATTATTCTG[A/G]GAAGGCCTATCTTGCTCTGTTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605882 MIM: 611346 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRIP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BRIP1 - BRCA1 interacting protein C-terminal helicase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
INTS2 - integrator complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020748.2 | 2798 | Silent Mutation | TCC,TCT | S,S 905 | NP_065799.1 | |
XM_006721987.3 | 2798 | Silent Mutation | TCC,TCT | S,S 897 | XP_006722050.1 | |
XM_011525058.2 | 2798 | Silent Mutation | TCC,TCT | S,S 897 | XP_011523360.1 |