Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCCGCCACCAGCCTCCGGCAG[G/T]CCCAGGCCATCTGCAGGGAAAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142967 MIM: 142966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOXB-AS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HOXB-AS1 - HOXB cluster antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HOXB2 - homeobox B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002145.3 | 522 | Silent Mutation | GGA,GGC | G,G 134 | NP_002136.1 | |
XM_005257275.4 | 522 | Silent Mutation | GGA,GGC | G,G 25 | XP_005257332.1 |
HOXB3 - homeobox B3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |