Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATCTTCATCTGAATCCAAATCC[A/G]TTGTGAATTTTGTTTTTGCTAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126430 | ||||||||||||||||||||
Literature Links: |
TOP2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOP2A - topoisomerase (DNA) II alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001067.3 | 4084 | Missense Mutation | ACG,ATG | T,M 1328 | NP_001058.2 | |
XM_005257632.1 | 4084 | Missense Mutation | ACG,ATG | T,M 1316 | XP_005257689.1 | |
XM_011525165.1 | 4084 | Missense Mutation | ACG,ATG | T,M 1328 | XP_011523467.1 |