Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCTGGTCCTTCAACAGAGGAAGG[A/G]TCCCCTTCTCCAGGGCCTCAGCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611221 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSDMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSDMB - gasdermin B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042471.1 | 3098 | Missense Mutation | ACC,ATC | T,I 346 | NP_001035936.1 | |
NM_001165958.1 | 3098 | Missense Mutation | ACC,ATC | T,I 359 | NP_001159430.1 | |
NM_001165959.1 | 3098 | Missense Mutation | ACC,ATC | T,I 350 | NP_001159431.1 | |
NM_018530.2 | 3098 | Missense Mutation | ACC,ATC | T,I 337 | NP_061000.2 | |
XM_011525004.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523306.1 | |
XM_011525006.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523308.1 | |
XM_011525007.2 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523309.1 | |
XM_011525009.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523311.1 | |
XM_011525011.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523313.1 | |
XM_011525012.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523314.1 | |
XM_011525013.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523315.1 | |
XM_011525015.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_011523317.1 | |
XM_017024848.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_016880337.1 | |
XM_017024849.1 | 3098 | Missense Mutation | ACC,ATC | T,I 363 | XP_016880338.1 | |
XM_017024850.1 | 3098 | Missense Mutation | ACC,ATC | T,I 359 | XP_016880339.1 | |
XM_017024851.1 | 3098 | Missense Mutation | ACC,ATC | T,I 354 | XP_016880340.1 | |
XM_017024852.1 | 3098 | Missense Mutation | ACC,ATC | T,I 346 | XP_016880341.1 | |
XM_017024853.1 | 3098 | Intron | XP_016880342.1 | |||
XM_017024854.1 | 3098 | Intron | XP_016880343.1 |