Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGCCTCCTATCTCAAGCTGTGAT[A/G]CAGGGTAGGTCATGAGCGCAGTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 156490 | ||||||||||||||||||||
Literature Links: |
NME1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NME1 - NME/NM23 nucleoside diphosphate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000269.2 | 320 | Intron | NP_000260.1 | |||
NM_198175.1 | 320 | Missense Mutation | ACA,GCA | T,A 23 | NP_937818.1 |
NME1-NME2 - NME1-NME2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018136.2 | 320 | Intron | NP_001018146.1 |