Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609575 MIM: 602887 MIM: 602151 MIM: 612910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACADVL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACADVL - acyl-CoA dehydrogenase, very long chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000018.3 | 1725 | Intron | NP_000009.1 | |||
NM_001033859.2 | 1725 | Intron | NP_001029031.1 | |||
NM_001270447.1 | 1725 | Intron | NP_001257376.1 | |||
NM_001270448.1 | 1725 | Intron | NP_001257377.1 | |||
XM_006721516.2 | 1725 | Intron | XP_006721579.2 | |||
XM_011523829.1 | 1725 | Intron | XP_011522131.1 | |||
XM_011523830.1 | 1725 | Intron | XP_011522132.1 |
DLG4 - discs large MAGUK scaffold protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DVL2 - dishevelled segment polarity protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004422.2 | 1725 | Silent Mutation | CCC,CCG | P,P 699 | NP_004413.1 | |
XM_005256502.1 | 1725 | Silent Mutation | CCC,CCG | P,P 695 | XP_005256559.1 | |
XM_017024298.1 | 1725 | Silent Mutation | CCC,CCG | P,P 439 | XP_016879787.1 |
MIR324 - microRNA 324 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHF23 - PHD finger protein 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |