Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGATGATGCGATCCTCGTCATCCT[C/T]GTCCTTGATGGCCGAGGCCATGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610786 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SRCIN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SRCIN1 - SRC kinase signaling inhibitor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025248.2 | 3460 | Missense Mutation | AAG,GAG | K,E 1079 | NP_079524.2 | |
XM_017025169.1 | 3460 | Missense Mutation | AAG,GAG | K,E 1115 | XP_016880658.1 | |
XM_017025170.1 | 3460 | Missense Mutation | AAG,GAG | K,E 1147 | XP_016880659.1 | |
XM_017025171.1 | 3460 | Missense Mutation | AAG,GAG | K,E 1147 | XP_016880660.1 | |
XM_017025172.1 | 3460 | Missense Mutation | AAG,GAG | K,E 1113 | XP_016880661.1 | |
XM_017025173.1 | 3460 | Missense Mutation | AAG,GAG | K,E 1147 | XP_016880662.1 | |
XM_017025174.1 | 3460 | Missense Mutation | AAG,GAG | K,E 933 | XP_016880663.1 |