Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTGCCCTTGACCTCAGCTACGA[C/T]GGAGGGCTTGGCTGAGGGGCGTGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603781 | ||||||||||||||||||||
Literature Links: |
MYO15B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MYO15B - myosin XVB | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RECQL5 - RecQ like helicase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003715.3 | 1739 | Intron | NP_001003715.1 | |||
NM_001003716.3 | 1739 | Intron | NP_001003716.1 | |||
NM_004259.6 | 1739 | Missense Mutation | NP_004250.4 | |||
XM_005257818.3 | 1739 | Missense Mutation | XP_005257875.1 | |||
XM_005257822.3 | 1739 | Missense Mutation | XP_005257879.1 | |||
XM_005257823.3 | 1739 | Intron | XP_005257880.1 | |||
XM_006722186.2 | 1739 | Missense Mutation | XP_006722249.1 | |||
XM_011525482.2 | 1739 | Missense Mutation | XP_011523784.1 | |||
XM_011525484.1 | 1739 | Missense Mutation | XP_011523786.1 | |||
XM_011525485.2 | 1739 | Missense Mutation | XP_011523787.1 | |||
XM_011525486.2 | 1739 | Missense Mutation | XP_011523788.1 | |||
XM_017025343.1 | 1739 | Missense Mutation | XP_016880832.1 | |||
XM_017025344.1 | 1739 | Intron | XP_016880833.1 |
SMIM5 - small integral membrane protein 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |