Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCAGTTGTTAAACCTAAAACAG[A/C]AAAAGAAAAGCTGAAGGAACTCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600396 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX8 - DEAH-box helicase 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302623.1 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_001289552.1 | |
NM_001322216.1 | 453 | UTR 5 | NP_001309145.1 | |||
NM_001322217.1 | 453 | Missense Mutation | GAA,GCA | E,A 20 | NP_001309146.1 | |
NM_001322218.1 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_001309147.1 | |
NM_001322219.1 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_001309148.1 | |
NM_001322220.1 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_001309149.1 | |
NM_001322221.1 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_001309150.1 | |
NM_004941.2 | 453 | Missense Mutation | GAA,GCA | E,A 111 | NP_004932.1 |