Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCCAATTGAAATTAGTAGCGA[C/T]GATAGCAAAGAAGACTATAGTTTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609534 MIM: 614853 | ||||||||||||||||||||
Literature Links: |
ATAD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD5 - ATPase family, AAA domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024857.4 | 816 | Silent Mutation | GAC,GAT | D,D 139 | NP_079133.3 | |
XM_011525269.2 | 816 | Silent Mutation | GAC,GAT | D,D 139 | XP_011523571.1 | |
XM_011525270.2 | 816 | Silent Mutation | GAC,GAT | D,D 139 | XP_011523572.1 | |
XM_011525273.2 | 816 | Silent Mutation | GAC,GAT | D,D 139 | XP_011523575.1 | |
XM_011525274.2 | 816 | Intron | XP_011523576.1 | |||
XM_017025111.1 | 816 | Silent Mutation | GAC,GAT | D,D 125 | XP_016880600.1 |
CRLF3 - cytokine receptor like factor 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |