Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTAAATAGACTGATGATGAGTA[C/T]ATTCCTCTCATTAAACTCCAACGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609534 MIM: 616422 | ||||||||||||||||||||
Literature Links: |
ATAD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD5 - ATPase family, AAA domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEFM - transcription elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024683.3 | 1172 | Missense Mutation | ATA,ATG | I,M 206 | NP_078959.3 | |
XM_006722084.1 | 1172 | Missense Mutation | ATA,ATG | I,M 91 | XP_006722147.1 |