Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTGGTGGTGGCGGTGGCGGCTG[C/G]GCTGGCTGCTGCTGTTGCTGTGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605523 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TOB1 - transducer of ERBB2, 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243877.1 | 981 | Silent Mutation | GCC,GCG | A,A 247 | NP_001230806.1 | |
NM_001243885.1 | 981 | Silent Mutation | GCC,GCG | A,A 108 | NP_001230814.1 | |
NM_005749.3 | 981 | Silent Mutation | GCC,GCG | A,A 247 | NP_005740.1 |
TOB1-AS1 - TOB1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |