Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCATCCAGTGCTCGATCAGCATGT[C/T]GGAACACGAGGTGGGTCCCTGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605483 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAI2 - dynein axonemal intermediate chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172810.1 | 300 | Missense Mutation | TCG,TTG | S,L 58 | NP_001166281.1 | |
NM_023036.4 | 300 | Missense Mutation | TCG,TTG | S,L 58 | NP_075462.3 | |
XM_011525125.2 | 300 | Missense Mutation | TCG,TTG | S,L 58 | XP_011523427.1 |