Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGACGTTCTCCCCTTCAATTCAA[C/T]GGGAAGGCTCTTTTCTGGCATGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194480 MIM: 616463 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928000 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101928000 - uncharacterized LOC101928000 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFP3 - ZFP3 zinc finger protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF232 - zinc finger protein 232 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320952.1 | 1449 | Missense Mutation | CAT,CGT | H,R 435 | NP_001307881.1 | |
NM_001320953.1 | 1449 | Missense Mutation | CAT,CGT | H,R 417 | NP_001307882.1 | |
NM_001320954.1 | 1449 | Missense Mutation | CAT,CGT | H,R 417 | NP_001307883.1 | |
NM_001320955.1 | 1449 | Missense Mutation | CAT,CGT | H,R 416 | NP_001307884.1 | |
NM_014519.3 | 1449 | Missense Mutation | CAT,CGT | H,R 444 | NP_055334.2 | |
XM_011524006.2 | 1449 | Missense Mutation | CAT,CGT | H,R 396 | XP_011522308.1 | |
XM_017025021.1 | 1449 | Missense Mutation | CAT,CGT | H,R 444 | XP_016880510.1 | |
XM_017025022.1 | 1449 | Missense Mutation | CAT,CGT | H,R 368 | XP_016880511.1 | |
XM_017025023.1 | 1449 | Missense Mutation | CAT,CGT | H,R 359 | XP_016880512.1 |