Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAAGCCACTCTTCCGCCACTTCC[A/G]CCGGATAGACTCCTGTCTGCAGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR6867 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6867 - microRNA 6867 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAPGEFL1 - Rap guanine nucleotide exchange factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303533.1 | 153 | Missense Mutation | CAC,CGC | H,R 147 | NP_001290462.1 | |
NM_001303534.1 | 153 | Missense Mutation | CAC,CGC | H,R 141 | NP_001290463.1 | |
NM_016339.4 | 153 | Missense Mutation | CAC,CGC | H,R 92 | NP_057423.1 | |
XM_006721932.3 | 153 | Missense Mutation | CAC,CGC | H,R 298 | XP_006721995.1 | |
XM_006721935.3 | 153 | Missense Mutation | CAC,CGC | H,R 129 | XP_006721998.1 | |
XM_006721937.3 | 153 | Missense Mutation | CAC,CGC | H,R 92 | XP_006722000.1 | |
XM_006721938.3 | 153 | Missense Mutation | CAC,CGC | H,R 28 | XP_006722001.1 |